Anti-PLPPR2 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A62654-100UG A62654-50UG
77720-643EA 529.22 CAD
77720-643 77720-644
Anti-PLPPR2 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PLPPR2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR