Luer Stubs (Blunt Needles)
Supplier: INSTECH LABORATORIES, INC
LS14 LS15 LS16 LS17 LS18 LS19 LS20 LS21 LS22 LS22/6 LS23 LS25 LS27
77679-320BX 142.45 CAD
77679-320 77679-322 77679-324 77679-326 77679-328 77679-330 77679-332 77679-334 77679-336 77679-338 77679-340 77679-342 77679-344
Luer Stubs (Blunt Needles)
Cannulae
Use these female luer stubs to connect infusion tubing or catheters to syringes or other male Luer connectors. The hubs are polypropylene and the tips are stainless steel. The tips are deburred to prevent damage to tubing.
  • Female Luers
  • Polypropylene hubs
  • Stainless steel tips
  • Deburred tips
Blunt tip needles (Luer stub adaptors) have numerous laboratory applications. When dosing, sampling, or infusing, these blunt needles ensure a secure and reliable connection to in-dwelling catheters. They provide a smooth, burr-free needle edge which preventsMore Product Information