Saturix C30L Lightweight Polyester/Cellulose Wipes
Supplier: FG CLEAN WIPES TE
7-C30-BBP-00 7-C30-99P-00
77695-986CS 528.91 CAD
77695-986 77695-984
Saturix C30L Lightweight Polyester/Cellulose Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These lightweight polyester/cellulose cleanroom wipes offer the essential benefits of non-woven wipes in a versatile, economical package. Exacting manufacturing processes enhance cleanliness for a wide range of applications.
  • Lightweight substrate
  • Hydroentangled
  • 55% cellulose/45% polyester
  • Absorbent
  • Extremely low particle generation
  • No chemical binders
  • Economical
  • Low NVRs and ions
FG Cleanwipes C30L products are engineered to meet the stringent requirements of cleanroom environments, offering superior strength, cleanliness, and versatility through their advanced polyester/cellulose blend. Treated to minimize ions, non-volatile residues (NVRs), and particulates, these wipes provide outstanding lint-free performance, ensuring optimal cleanliness for sensitive applications.
Their versatility makes them perfect for cleaning critical surfaces in controlled environments. Offering enhanced cleanliness, they feature a special treatment that ensures minimal particulate release, maintaining optimal cleanliness. Their design combines strength and durability with softness for precision cleaning without surface damage.
Targeted industries and customers:
Pharmaceutical and Biotech: Essential for maintaining sterile environments and cleanroom operations.
Electronics: Ideal for contamination-free environments in semiconductor and electronics manufacturing.
Medical device manufacturing: supports stringent cleanliness standards, providing precision cleaning for compliance.
C30L wipes are a vital tool for industries prioritizing contamination control, safety, and operational efficiency.