Saturix C3 Hydroentangled 100% Polyester Wipes
Supplier: FG CLEAN WIPES TE
7-C3-44L-00 7-C3-99L-00 7-C3-BBL-00
77695-988CS 1826.48 CAD
77695-988 77695-990 77695-992
Saturix C3 Hydroentangled 100% Polyester Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These 100% hydroentangled polyester cleanroom wipes offer the highest level of softness and abrasion resistance; perfect for cleaning and polishing the most sensitive surfaces. But that softness does not come with a trade-off for durability. These cleanroom wipes are built tough for use in a variety of applications.
  • Comprised of 100% hydroentangled polyester fibers
  • Absorbent
  • No chemical binders
  • Recommended for the cleaning of lenses or other sensitive surfaces
  • Chemically compatible with most cleaning solutions
  • Pure and clean
  • Low NVRs and ions
FG Cleanwipes C3 products are designed to meet the critical demands of cleanroom environments, offering superior cleanliness, strength, and versatility through their advanced polyester/cellulose blend. Treated to minimize ions, non-volatile residues (NVRs), and particulates, these wipes deliver exceptional lint-free performance for contamination-sensitive applications.
Their versatility makes them ideal for cleaning critical surfaces in controlled environments. Offering enhanced cleanliness, they feature a special treatment that ensures minimal particulate release, maintaining optimal cleanliness. Their design combines strength and durability with softness for precision cleaning without surface damage.
Targeted industries and customers:
Pharmaceutical and Biotech: Essential for sterile manufacturing and cleanroom maintenance.
Electronics: Ensures contamination-free environments in semiconductor and electronics assembly.
Medical device manufacturing: Supports compliance with stringent cleanliness standards, enabling precise cleaning.
C3 wipes are indispensable for industries focused on contamination control, safety, and efficiency.