VWR® 900 Portable Conductivity Meters
Supplier: SCHULER SCIENTIFIC
VWR-EC900 77618-982 VWR-EC900B VWR-EC900C
77618-980EA 1863.99 CAD
77618-980 77618-982 77618-984 77618-986
VWR® 900 Portable Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC900 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature in any environment.
  • High resolution color touch display (4.3")
  • Five-point calibration with standard recognition
  • IP65 waterproof
  • Handheld
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 1000 data sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
The easy to use VWR-EC900 portable meters provide accurate conductivity measurements inside and outside the lab. The intelligent operation system provides features including user managment, calibration management, electrode management, method management, and many other advanced capabilities.
Ordering information: Three-year meter warranty. For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
Delivery information: All meters include carrying case, electrode holders (2), meter silicone rubber case, touch screen pen, and hand strap.