MiniTex Fume Extractor Arms, Movex
Supplier: Movex
MXT1100-75 MXT1100-100 MXT1500-75 MXT1500-100 MTI500-125 MTI750-125 MTI1000-125 MVK-125
89038-698EA 1776.73 CAD
89038-698 89038-700 89038-702 89038-704 89038-706 89038-708 89038-710 89038-712 89038-720 89038-724 89038-728 89038-732
MiniTex Fume Extractor Arms, Movex
Fume Extractors
Flexible, telescopic fume extractor arms for laboratory applications that require efficient operation when available pressure in the fan system is low
Self-adjustable telescopic function, movable angle joint, and 360° rotation ensure that the arm stays in position when adjusted. Flexible hose is made of spring steel coil completely embedded in PVC-coated, nonflammable polyamide fabric. Supporting brackets are mounted outside the hose to ensure natural, undisturbed airflow with minimal pressure drop and duct blockage. Maintenance and joint adjustment can be performed from the outside withMore Product Information