R and RX Fume Extractor Arms, Movex
Supplier: Avantor
89038-570EA 2333.66 CAD
89038-570 89038-572 89038-578 89038-584 89038-586 89038-588 89038-590 89038-592 89038-594
R and RX Fume Extractor Arms, Movex
Fume Extractors
Stable construction with a gas spring that balances the dead weight of the arm. Suitable for welding fumes or flue gas.
  • Flexible hose is made of spring steel coil completely embedded in PVC-coated, nonflammable polyamide fabric
  • Supporting brackets are mounted outside the hose to ensure natural, undisturbed airflow with minimal pressure drop and duct blockage
  • Maintenance and joint adjustment can be performed from the outside with hand tools
The RX model fume extractor arms have straight sections made of powder-coated, rounded sheet steel. These RX units are also provided with a flexible suction nozzle that can be rotated 360° and moved to a 90° angle.
Operating temperature range for all models: –10 to 90°C (14 to 194°F).
Color: Blue with black
Ordering information: Fume extractor arms are supplied fully assembled with a bracket for mounting on a wall. Conical hoods 89038-596 and 89038-598 come with a 12VAC, 50W halogen light. For white or stainless steel units, contact your Avantor sales representative.