Benchtop Extraction Arms, Nederman
Supplier: Nederman
70510144 70510244 70510444 70510344 70510544 70540144 70540344 70540244 70540444 70570144 70570244 70570344 70530144 70530244 70530344 70530444 70530544 70560144 70560344 70560444 70550144 70550344 70580144 70580244 70580344 70580544 70502834
CA89087-702EA 701.44 CAD
CA89087-702 CA89087-704 CA89087-706 CA89087-708 CA89087-710 CA89087-712 CA89087-714 CA89087-716 CA89087-718 CA89087-720 CA89087-722 CA89087-724 CA89087-728 CA89087-730 CA89087-732 CA89087-734 CA89087-736 CA89087-738 CA89087-740 CA89087-744 CA89087-746 CA89087-748 CA89087-754 CA89087-756 CA89087-758 CA89087-760 CA89087-832
Benchtop Extraction Arms, Nederman
Fume Extractors
Ideal for applications where small, flexible, and easy-to-use arms are required to eliminate smoke, gases, dust, and particles
The Original model is a robust and reliable arm for general applications with little risk of electrical discharges or attack by aggressive chemicals. The ESD/EX model is designed for use in electronic and explosive environments with risk of electrostatic discharges. ESD/EX model arms are made of conductive materials, eliminating the risk of static electricity and sparks. The Chem model has profiles made of double anodized aluminum and is ideal for fume extractionMore Product Information