Canopy Hood, Labconco®
Supplier: Labconco
4799000 7099000
30192-104EA 5822.3 CAD
30192-104 30192-159
Canopy Hood, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Specifically designed to solve unique ventilation problems, this canopy hood vents nontoxic materials such as heat, steam, and odors from large or bulky equipment.
Hood is shipped fully assembled with integral baffle and mounting hardware for peninsular or island locations*. Hood interior and exterior are epoxy-coated, cold-rolled steel. Requires remotely located blower for operation. Contact your Avantor Furniture Specialist for assistance with blower sizing.
*For ceiling mount, threaded rods (not included) may be required.
The epoxy-coated steel canopy has a built-in baffle toMore Product Information