REDISHIP Auxiliary Air Chambers, Kewaunee®
Supplier: Kewaunee
CF5-1372-03 CF5-1348-03
CF5-1348-03EA 4632.15 CAD
CF5-1348-03 CF5-1372-03
REDISHIP Auxiliary Air Chambers, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
These chambers can be added to the top of any Supreme Air fume hood to convert it to an auxiliary air fume hood.
  • Includes all necessary parts except a fan and ductwork
  • Requires no modifications to the fume hood
  • Chambers are made of cold-rolled steel
  • Finish is light neutral color or snow white
Adding the chamber allows the fume hood to operate on up to 70% auxiliary air, saving expensive, conditioned room air from being exhausted.
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order.