REDISHIP Supreme Air® Fume Hood Worktops, Kewaunee®
Supplier: Kewaunee
5-0204-A1 5-0204-A8 5-0205-A1 5-0205-A8 5-0206-A1 5-0206-A8 5-0208-A1 5-0208-A8
VF5-0204-A1EA 3775.19 CAD
VF5-0204-A1 VF5-0204-A8 VF5-0205-A1 VF5-0205-A8 VF5-0206-A1 VF5-0206-A8 VF5-0208-A1 VF5-0208-A8
REDISHIP Supreme Air® Fume Hood Worktops, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
These modified epoxy resin worktops are available with or without a 5.1cm (2") diameter vent hole
Use the vent hole for vented acid storage base cabinets. The 1.8m (6') and 2.4m (8') models require two base cabinets (vent is provided only for the right-side cabinet). The 2.4m (8') worktop comes in two pieces
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order.