REDISHIP Supreme Air® Front Load Fittings for Isotope Bench Hoods, Kewaunee®
Supplier: WaterSaver Faucet
L739LWPHLRTL L739LNPHLRP L739RWPHLRTL L739RNPHLRP L739LWPHLRP L739RWPHLRP
VFT-0739-L-FDEA 726.58 CAD
VFT-0739-L-FD VFT-0739-L-NIT VFT-0739-R-FD VFT-0739-R-NIT VFT-0739-L-CW VFT-0739-R-CW
REDISHIP Supreme Air® Front Load Fittings for Isotope Bench Hoods, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
Since isotope hoods do not have interior access panels, they require a front load fitting that can be accessed from outside the hood
Front load fittings will not fit on standard KMER-lined bench or walk-in hoods.
Ordering information: REDISHIP units can be shipped within 48 hours of receiving your order. Fittings include brass valve with replaceable seat and nylon color-coded panel flange with angle serrated hose connector.