SafeMask® Premier Elite™ ProShield Ear Loop Masks with Visor, ASTM Level 3
Supplier: Medicom
204215 204216 204216
CA77705-812BX 30.36 CAD
CA77705-812 CA77705-814 77705-814
SafeMask® Premier Elite™ ProShield Ear Loop Masks with Visor, ASTM Level 3
Masks Hygiene Masks
ASTM Level 3 mask with integrated visor ensures full face protection.
  • ≥98% bacterial and particle filtration efficiency, providing the highest level of procedure mask protection
  • Fluid-resistant mask construction plus unique shingle pleat design prevents fluid pooling and cross-contamination
  • Anti-fog, distortion-free eye shield ensures clear vision
  • Latex-free adjustable nosepiece and soft, strong ear loops deliver a comfortable and customized fit
  • Made from premium materials, mask will not lint, tear or shred
When applications require more protection than a mask alone can provide, choose the SafeMask® Premier Elite™ ProShield. According to the CDC, protective eyewear with gaps between glasses and the face likely do not protect eyes from all splashes and sprays.
Premier Elite™ ProShield is a high-barrier mask with an integrated wrap-around shield that reliably protects the eyes, making it ideal for treating high-risk patients and for procedures producing heavy amounts of airborne particles, moisture, spray, and spatter.
Material components include Spunbond Polypropylene (SBPP), Melt Blown Polypropylene (MBPP), aluminum, and polyester.
Certifications: ASTM F2100 Level 3 barrier protection.
Packaging: Masks are packaged 25 per box, 4 boxes per case (100 masks per case).