VWR® 900 Portable Dissolved Oxygen Meters
Supplier: SCHULER SCIENTIFIC
VWR-DO900 VWR-DO900A
77618-936EA 1843.78 CAD
77618-936 77618-938
VWR® 900 Portable Dissolved Oxygen Meters
Electrochemistry Meters Oxygen Meters
The VWR-DO900 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
  • High resolution color touch display (4.3")
  • Air-saturated water or zero oxygen calibration capabilities
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 1000 sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Auto barometric pressure compensation
The easy to use VWR-DO900 portable meters provide accurate dissolved oxygen measurements for accurate water quality testing. The intelligent operation system provides features including user managment, calibration management, electrode management, method management, and many other advanced capabilities.
Ordering information: For use with DO1 polarographic DO electrode (77618-816). Three-year meter warranty. For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the related information section.
Delivery information: All meters include electrode holders (2), meter silicone rubber case, carrying case, touch screen pen and hand strap.