VWR® 900 Portable Ion/pH Meters
Supplier: VWR International
77619-030 77619-032 77619-034 77619-036 77618-922 77619-050 77619-052 77619-054 77619-056 77619-058 77619-028 77619-038 77619-040 77619-042 77619-044 77619-046 77619-048
77619-030EA 2611.6 CAD
77619-030 77619-032 77619-034 77619-036 77618-922 77619-050 77619-052 77619-054 77619-056 77619-058 77619-028 77619-038 77619-040 77619-042 77619-044 77619-046 77619-048
VWR® 900 Portable Ion/pH Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-I900 meter provide accurate and reliable measurements of ISE, pH, mV and temperature in any environment.
  • High resolution color touchscreen (4.3")
  • 1 to 8 point calibration with standard recognition for pH
  • Built-in ion methods including F⁻, CL⁻, K⁺, and Na⁺
  • Selectable pH buffer groups including NIST, DIN, GB, USA
  • IP 65 waterproof
  • Ability to save 1000 sets of measurement results (GLP-compliant)
  • Rechargeable battery
  • Three-year meter warranty
Automatic or Manual temperature compensation capabilities. Auto-read, timed-read, and continuous-read measurement options. Automatic electrode diagnosis with pH slope and offset display.
The easy to use VWR-I900 portable meters provide accurate measurements in the field and the laboratory. The intelligent operation system provides features including user management, calibration management, electrode management, method management, and many other advanced capabilities. The included carrying case allows for easy transport of all items to make measurements on the go even easier.
For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the related information section.
Delivery information: All meters supplied with: electrode holders (2), meter silicone rubber case, carrying case, touch screen pen and hand strap.