Volkman Bone Curette
Supplier: WORLD PRECISION INSTRUMENTS LLC
503752 503753
77663-416EA 77.89 CAD
77663-416 77663-418
Volkman Bone Curette
Curettes
The Volkman bone curette is a bone curette for debulking, and it has a razor-sharp cutting edge.
  • It is commonly used in orthopedic procedures
  • The knurled round handle of this curette gives you a sure grip
  • The handle gradually tapers to the tips
Removal of fibrous tissue from bone or scrape bone to remove small pieces or shape it with this curette. This cutting edge is an oval scoop at the end of the curette. This is manufactured for surgical grade stainless steel and is suitable for sterilization via autoclaving or chemical sterilant.