Mayo Hegar Needle Holders, Straight, Disposable
Supplier: WORLD PRECISION INSTRUMENTS LLC
503732-12 503733-12
77663-410PK 79 CAD
77663-410 77663-412
Mayo Hegar Needle Holders, Straight, Disposable
Forceps
Mayo-Hegar needle holders are surgical instruments designed for holding and driving needles through tissues.
  • Mayo-Hegar needle holders are designed with a scissor-like shape
  • Featuring a long
  • Slender body with finger rings on one end and opposed jaws on the other
The jaws have serrated (cross-hatched) tips to securely grip the needle during suturing. They feature a ratcheted locking mechanism. These needle holders are 15 cm (6") long, and the tips are straight. These are made of surgical grade stainless steel, suitable for autoclaving and other sterilization methods. This is a package of 12 instruments that are student grade instruments.