Hartman Mosquito Forceps, Disposable
Supplier: WORLD PRECISION INSTRUMENTS LLC
503728-12 503729-12
77663-402PK 87.6 CAD
77663-402 77663-404
Hartman Mosquito Forceps, Disposable
Forceps
Hartman Mosquito Forceps, also called Hartmann mosquito hemostatic forceps, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip.
  • Mosquito Forceps have long, thin, and straight or curved jaws
  • Jaws are serrated for a firm grasp with minimal tissue trauma
  • Handles of mosquito forceps have a ratchet lock to secure
They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Hartman Mosquito Forceps have a larger tip and deeper serrations than Halsted Mosquito Forceps. They are designed for clamping larger blood vessels.
Packaging: Forceps are sold in packages of 12.