KIMBLE® ULTRA-WARE® Microfiltration Apparatus with Solvent Pickup Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953820-1047 953820-4047
KT953820-1047EA 1977.52 CAD
KT953820-1047 KT953820-4047
KIMBLE® ULTRA-WARE® Microfiltration Apparatus with Solvent Pickup Adapter, DWK Life Sciences
Filters Bottle Top Filters
Eliminates the pouring of HPLC solvents by using a solvent pickup adapter to draw solvent directly from a reagent bottle through a 47 mm filter.
Features the Hi-Vac® valve on the solvent pickup adapter that can be turned off for vacuum degassing. Filter base has an integral hose connection above the filtrate drip tip to prevent contamination of the vacuum line with filtrate droplets.
Ordering information: Component parts may be ordered separately to convert existing ULTRA-WARE® and Millipore® filtration assemblies for in-line filtration/degassing.
Delivery information: System includes a solvent pickup adapter, coarse porosity fritted glass filter base, inner joint flask, and an aluminum clamp.