Flexible Duct Connections, Labconco
Supplier: Labconco
4861800 7034200 5621400 4726500
30152-642EA 360.24 CAD
30152-642 30182-500 30182-502 82006-956
Flexible Duct Connections, Labconco
Ducting Supplies
Flexible duct connections reduce vibration between the blower and PVC ductwork.
Supplied with two clamps for easy installation.
Order Now
SPECIFICATIONS
Environmentally Preferable