Microinjection Syringe Pumps
Supplier: WORLD PRECISION INSTRUMENTS LLC
UMP3T-2 UMP3T-1
77665-476EA 9702.78 CAD
77665-476 77665-474
Microinjection Syringe Pumps
Pumps Syringe Pumps
The UMP3 (UltraMicroPump3) is a versatile microinjection syringe pump paired with microsyringes to deliver microliter-level volumes. The UMP3 is optimal for applications requiring precise, targeted injections of extremely low-volume samples. With its touchscreen controller, our UMP3 can deliver or withdraw as little as 25 nl (using 10 µl syringe with 60 mm stroke length).
  • Accepts a wide range of microinjection syringes from 0.5 to 1000 µl
  • Manual or automated functionality
  • Smooth and low-noise operation
  • Mounts directly on micromanipulator or stereotaxic frame
  • Injections down to 25 nl
  • Rapid setup with intuitive touchscreen controller
Applications: Accurate microinjection of solution into tissue, cannulation drug delivery, micro delivery of biochemical agents or dyes, intravitreal injection and intraoccular injection. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps.