Multifunction Microforge Controllers
Supplier: WORLD PRECISION INSTRUMENTS LLC
DMF1000-1 DMF1000-2 DMF1000-M1 DMF1000-M2
77664-204EA 10767.7 CAD
77664-204 77664-206 77664-210 77664-212
Multifunction Microforge Controllers
Microinjectors Microinjectors, Programmable
The DMF1000 is a microprocessor-controlled microforge offering unmatched performance. Designed for fabrication of both small patch clamp glass pipettes and larger injection pipettes, the DMF1000 should find many uses in the laboratory. The extensive improvements incorporated into the DMF1000 greatly increase its versatility and performance, making it one of the most powerful microforges on the market.
  • Microforging, micropipette calibration and microinjection - in a single device
  • Microprocessor-controlled microforge
  • Digital signal processor technology precisely controls the polish heating time
  • Unique digital pneumatic pressure feature polishes the tip without changing the size
  • Shipping weight: 7.3 kg
  • Dimensions: 10.2×17.8×4.8 cm
  • Power: 100 to 240 VAC 50/60 Hz
Benefits:
Digital Signal Processor (DSP) technology
Complete system package available
Kohler illuminator and Abbe condenser for less glare and sharper images.
Pneumatic pressure polishing that allows the preparation of blunt tips without change of tip ID
Heating filament is attached to the microscope objective so they move together
Pipette holder sits on the microscope stage to simplify the locating and polishing of the pipette
Applications:
Polishing patch pipettes
Microforging holding pipettes
Microforging beveled injection pipettes
Pipette tip calibration and microinjection
Digital Signal Processor (DSP) Technology: The DMF1000 is powered by the latest digital signal processor (DSP) technology. A digital timer is used to precisely control the polish heating time. Ten memories can be used to store settings of the heating power and heating duration. All of the settings are controlled and displayed digitally for better accuracy and reproducibility. Two different operating modes are provided: Manual and Auto. In the Manual mode, the DSP will memorize the duration of the time that is used to achieve a desired polishing. In Auto mode, the heat will be applied for the duration of the timer setting.
Kohler Illuminator and Abbe Condenser: Other benefits of the DMF1000 design include the use of a Kohler illuminator and Abbe condenser, which provide the reduced glare and sharper image contrast necessary when polishing pipettes as small as half a micron (0.5 µm) in diameter.
Pressure Polishing: The DMF1000 incorporates a unique digital pneumatic pressure feature that enables pressurized air to be delivered through the pipette during fire polishing. In the fabrication of patch pipettes, the pressurized air can be used to blunt the taper at the pipette tip without changing the size of the tip opening. This reduces electrical resistance of the tip, leading to lower noise during patch-clamp recordings.
Complete System: The DMF1000 system includes a specially configured WPI model W30S-LED research grade compound microscope (optional) equipped with a high quality metallurgic 40× long-working distance objective and a pair of 10× eyepieces. The long working distance objective reduces the danger of damage to the objective lens during the heating process.