Thermoplastic Ducts, Labconco
Supplier: Labconco
4708600 7027200 5602000 5605000 4718900
30152-636EA 1082.3 CAD
30152-636 30182-268 30182-504 82006-918 30182-086
Thermoplastic Ducts, Labconco
Ducting Supplies
PVC exhaust duct is Type 1, unplasticized, schedule 40, lightweight, and corrosion-resistant
A female duct coupling is required to join two sections. Connections can be made using solvent cement. PVC is rigid and may be cut without the use of special tools