HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99.0%

Supplier: IBI Scientific

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

IB01131 IB01132 IB01133
IB01131EA 184.86 CAD
IB01131 IB01132 IB01133
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99.0%
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid


  • For use in polyacrylamide electrophoresis buffer systems
  • Idealcell culture buffer
  • Maintains physiological pH in cell culture applications


Ideally suited for cell culture in a range of 10–25 mM. The zwitterionic buffer has a buffering ranges of 6.8–8.2. It can also be used as a component of TRIS-HEPES buffer for polyacrylamide electrophoresis of proteins. DNase-, RNase-, and protease-free.


Caution: Store at room temperature. Protect from moisture. Irritant. Avoid contact with eyes, skin, and mucous membranes. Avoid breathing vapors. Wash thoroughly with water after handling.

Formula: C₈H₁₈N₂O₄S
MDL Number: MFCD00006158
CAS Number: 7365-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline Powder
pH (0.5 M solution in H₂O): 5.1 to 6.5
Purity Min. 99.0
Water Max. 0.25%
Heavy Metals Max. 0.001%
Iron Max. 0.0005%
Sulfates Max. 0.05%
DNase assay None Detected
RNase assay None Detected
Protease assay None Detected
Cell Culture Testing Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR