LiChroCART® Stainless Steel Superspher® Cartridges, MilliporeSigma
Supplier: MilliporeSigma
1.16857.0001 1.50193.0001 1.50198.0001 1.50792.0001 1.50973.0001 1.50974.0001 1.50975.0001 1.50980.0001 1.51299.0001 1.16009.0001 1.16010.0001 1.16051.0001 1.16052.0001 1.16054.0001 1.16854.0001 1.16855.0001 1.16858.0001 1.16869.0001 1.16009.0001 1.16010.0001 1.16051.0001 1.16052.0001 1.16054.0001 1.16056.0001 1.16854.0001 1.16855.0001 1.16857.0001 1.16858.0001 1.16869.0001 1.50193.0001 1.50198.0001 1.50792.0001 1.50973.0001 1.50974.0001 1.50975.0001 1.51299.0001 1.50980.0001 1.50791.0001 1.51288.0001 1.51909.0001
48219-184EA 2828.42 CAD
48219-184 48219-526 48219-530 48219-550 48219-602 48219-604 48219-606 48219-610 48219-916 48219-150 48219-152 48219-164 48219-166 48219-168 48219-180 48219-182 48219-186 48219-190 CA48219-150 CA48219-152 CA48219-164 CA48219-166 CA48219-168 CA48219-170 CA48219-180 CA48219-182 CA48219-184 CA48219-186 CA48219-190 CA48219-526 CA48219-530 CA48219-550 CA48219-602 CA48219-604 CA48219-606 CA48219-916 EM1.50205.0001 CA1.50205.0001 CA48219-610 CA1.50791.0001 CA1.51288.0001 EM1.51909.0001
LiChroCART® Stainless Steel Superspher® Cartridges, MilliporeSigma
Chromatography Columns
These cartridges are ideal for highly efficient HPLC analysis of complex mixtures where high peak capacity is required, because the guaranteed number of theoretical plates for Superspher® cartridges is <100.000 N/m.
These LiChroCART® cartridges are compatible with manu-CART® '4' parts, which facilitates faster and more flexible method adaptation to smaller bore columns. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent.
This high performance spherical silica carrier yields anMore Product Information