Nalgene® Lower Assemblies/Shafts and Impellers, Thermo Scientific
Supplier: Thermo Fisher Scientific
2654-0030 2654-0055 2654-0100
66463-221CS 5903.96 CAD
66463-221 66463-223 66463-227
Nalgene® Lower Assemblies/Shafts and Impellers, Thermo Scientific
Stirrers Stirring Tools
Components are for use with BioTech Mixer Overhead Drive (see 66463-212 series)
Recommended vessel (sold separately) is listed below. Lower assemblies use type 316 stainless steel axial-flow impellers and shafts. All require sanitary mixer support (see 66463-210 series).