Hibar® LiChrosorb® and LiChrospher® HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.50316.0001 1.50318.0001 1.50329.0001 1.50332.0001 1.50333.0001 1.50432.0001 1.50433.0001 1.50377.0001 1.50388.0001 1.50477.0001 1.50333.0001 1.50316.0001 1.50318.0001 1.50329.0001 1.50332.0001 1.50377.0001 1.50388.0001 1.50432.0001 1.50433.0001 1.50477.0001 1.50545.0001 1.50546.0001 1.50547.0001 1.50549.0001 1.50550.0001 1.50573.0001 1.50574.0001 1.50575.0001 1.50578.0001 1.50579.0001 1.50580.0001 1.50581.0001 1.50582.0001 1.50583.0001 1.51903.0001
48219-322EA 549.69 CAD
48219-322 48219-324 48219-326 48219-328 48219-330 48219-347 48219-348 48219-340 48219-342 48219-354 CA1.50333.0001 CA48219-322 CA48219-324 CA48219-326 CA48219-328 CA48219-340 CA48219-342 CA48219-347 CA48219-348 CA48219-354 CA1.50545.0001 CA1.50546.0001 CA1.50547.0001 CA1.50548.0001 CA1.50549.0001 CA1.50550.0001 CA1.50573.0001 CA1.50574.0001 CA1.50575.0001 CA1.50578.0001 CA1.50579.0001 CA1.50580.0001 CA1.50581.0001 CA1.50582.0001 CA1.50583.0001 EM1.51903.0001
Hibar® LiChrosorb® and LiChrospher® HPLC Columns, MilliporeSigma
Chromatography Columns
For analytical HPLC.
RP-18 is reversed phase C18 (with an octadecylsilyl chain). RP-8 is reversed phase C8 (with an octylsilyl chain).
Cartridge holder 48219-176 is for the direct coupling of Hibar® columns. The holder replaces the original top fitting and holds any 4x4mm LiChroCART® guard column.
LiChrosorb® columns are available with 5µm or 10µm irregular media. LiChrospher® columns are available with 5 or 10 µm spherical media. Stainless steel columns are complete with fittings at both ends and shipped with an individual test chromatogram. May be connected to any HPLC instrument using standard 1.6mm (¹/₁₆") capillary male connections. To prolong the life of analytical columns, use with suitable precolumns.
Ordering information: Hibar® columns include end fittings.