Steri-Wrap® I, Propper Manufacturing
Supplier: Propper
Steri-Wrap®
31103620 31101200 31101500 31102400 311048
58752-920CS 807.03 CAD
58752-920 58752-863 58752-965 58752-861 89125-868
Steri-Wrap® I, Propper Manufacturing
Sterilization Wraps
Steri-Wrap® I is a medium weight wrap, ideal for general hospital use in steam and ethylene oxide sterilization cycles.
  • Cellulose based wrap for Steam and EO sterilization
  • Efficient microbial barrier, high-level of protection against fluids and particles
  • Highly permeable meaning no wet packs
  • Variety of strength of fabric to meet your needs
Steri-Wrap® I is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® I is a medium weight wrap, ideal for general hospital use. Steri-Wrap® I is designed to be virtually lint free with tightly anchored fibers. Wrap is reversible and features excellent liquid repellency.
Certifications: ISO 11607-1:2019