Autoclave Wraps, HPK Industries
Supplier: HPK Industries
083-RD 084-RD 085-RD 088-RD 087-RD 090-RD 089-RD 091-RD 086-RD
82004-130CS 1148.47 CAD
82004-130 82004-120 82004-132 82004-122 82004-134 82004-124 82004-126 82004-116 82004-128
Autoclave Wraps, HPK Industries
Sterilization Wraps
Use these autoclave wraps to sterilize apparel, instruments, trays, and more
Designed for use in steam, ethylene oxide, and gamma sterilization processes. Resistant to both aqueous-borne and airborne bacterial penetration. These Autoclave Wraps are constructed from SMS material; Spunbonded Polypropylene, Meltblown Polypropylene, Spunbonded Polypropylene composite. Wraps provide excellent barrier characteristics and are more environmentally friendly than plastic and/or synthetic substrates.