Gamma Radiation Sterilization Indicator Labels
Supplier: Crosstex
NMCPI-R01 NMCPI-R02 NMCPI-R03
19872-034EA 399.55 CAD
19872-034 19872-036 19872-038
Gamma Radiation Sterilization Indicator Labels
Sterilisation Indicators
Self-adhesive, nontoxic, chemical process indicators change from yellow to red upon exposure to gamma or electron beam radiation.
Labels are coated with a UV blocking agent for protection from exposure to UV light. A minimum of 15kGy (1.5Mrad) irradiation is recommended for sufficient endpoint color development. At greater dosage levels, the radiation indicators exhibit a darker shade of red. When exposed, imprinted indicators 19872-038 read “RED IS [gamma] EXPOSED.” Indicators are stable for at least 24 months from date of manufacture when they are stored between 15 to 30°C.
Certifications: Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.