Flexible Vent Assemblies, Scientific Plastics
Supplier: Scientific Plastics
F2738567-150 F160L078F-150
97002-254EA 397.92 CAD
97002-254 97002-256
Flexible Vent Assemblies, Scientific Plastics
Furniture Casework
Venting systems are designed for use in acid and solvent cabinets.
  • All vents are manufactured from acid-resistant polyolefin material
  • Non-squeezed top
The flexible tubing design allows systems to be installed around potential obstacles within existing cabinet structures. A 3 m (10') length of accessory tubing is available separately for field modifications.