Drum Dolly, Wesco
Supplier: Wesco
240030 240088
21002-018EA 479.45 CAD
21002-018 21002-020
Drum Dolly, Wesco
Trolleys
Welded steel frame with reinforced outriggers securely holds and transports 208.2 L (55gal.) or 322 L (85gal.) drums
  • Drum chime over caster kingpin provides stability and long caster life
  • Sintered iron casters measure 7.6 dia. x 3.2 W cm (3 x 11/4")
  • Capacity: 454 kg (1000 lbs.)