Pressure Tubes with Ace-Thred and Front Seal Plugs, Ace Glass
Supplier: Ace Glass
8648-04 8648-07 8648-09 8648-10 8648-102 8648-105 8648-109 8648-113 8648-115 8648-117 8648-12 8648-121 8648-123 8648-127 8648-136 8648-17 8648-21 8648-230 8648-232 8648-234 8648-245 8648-247 8648-249 8648-25 8648-251 8648-253 8648-308 8648-310 8648-312 8648-34 8648-36 8648-38 8648-43 8648-48 8648-53 8648-55 8648-77 8648-88 8648-96 8648-129
89063-162EA 225.59 CAD
89063-162 89063-166 89063-170 89063-172 89063-174 89063-180 89063-188 89063-194 89063-198 89063-202 89063-206 89063-210 89063-214 89063-222 89063-234 89063-260 89063-294 89063-298 89063-300 89063-302 89063-306 89063-310 89063-314 89063-316 89063-320 89063-324 89063-336 89063-340 89063-344 89063-352 89063-354 89063-356 89063-364 89063-372 89063-380 89063-382 89063-408 89063-422 89063-426 76318-586
Pressure Tubes with Ace-Thred and Front Seal Plugs, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are designed for polymerization and analysis of halogens by thermal decomposition in the presence of lime
Larger volume hydrogenation bottles have a flat bottom and a lower recommended operating pressure of 60psi. Bottle pressure ratings are determined at ambient temperature.
Tubes feature Ace-Thred closures which allow for quick and simple plug sealing and removal. Supplied with front seal plug and O-ring for pressure work. This closure system assures a tight and secure seal without the need for any extra equipment. Closures are reusable. Tubes areMore Product Information