Pressure Vessels, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1880-43 CG-1880-44 CG-1880-21 CG-1880-22 CG-1880-23 CG-1880-24 CG-1880-25 CG-1880-26 CG-1880-30 CG-1880-31 CG-1880-32 CG-1880-R-01 CG-1880-R-02 CG-1880-R-03 CG-1880-R-04 CG-1880-R-10 CG-1880-R-11 CG-1880-R-12 CG-1880-R-13 CG-1880-40 CG-1880-41 CG-1880-42 CG-1880-01 CG-1880-02 CG-1880-03 CG-1880-05 CG-1880-06 CG-1880-10 CG-1880-11 CG-1880-12 CG-1880-45 CG-1880-04
47733-812EA 308.73 CAD
47733-812 47733-814 80071-428 80071-430 80071-432 80071-434 80071-436 80071-438 80071-440 80071-442 80071-444 89028-802 89028-804 89028-806 89028-808 89028-810 89028-812 89028-814 89028-816 47733-806 47733-808 47733-810 80071-410 80071-412 80071-414 80071-418 80071-420 80071-422 80071-424 80071-426 10171-006 10171-008
Pressure Vessels, Heavy Wall, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy wall cylindrical and round bottom vessels have an internal thread for use with a 15 or 25mm fluoropolymer resin bushing as a pressure seal
This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime.
Ordering information: A complete apparatus is supplied with vessel, fluoropolymer resin bushing, and Viton® O-ring.