Harvard Trip Balances, Ohaus
Supplier: Ohaus
80000004 80000005 80000003
12350-032EA 948.85 CAD
12350-032 12351-024 12344-030 12353-008
Harvard Trip Balances, Ohaus
Balances and Scales
Versatile, rugged, easy-to-use balances are ideal for comparative weighing
  • 10 x 0.1 g (Upper Beam), 200 x 10 g (Lower Beam)
The footed polypropylene scoop with counterweight may be ordered separately.
Balances feature angled beam with center reading poise, magnetic damping, zero adjust compensator, and precision ground knives. Stainless steel 15.2cm (6") weighing platform. Balance 12351-024 includes a 225g tare beam.
Ordering information: Supplied with manufacturer’s five-year warranty.