Welch® Barbed Tubing Connectors
Supplier: Gardner Denver
3551K-14 3551K-13 3551K-12 3551K-51 3551K-90 3551K-05 3551K-02 3551K-04 3551K-01 3551K-06 3551K-03
84009-584PK 25.19 CAD
84009-584 84009-596 84009-574 84009-610 84009-598 84009-576 84009-600 84009-578 80094-366 84009-580 84009-582
Welch® Barbed Tubing Connectors
Fittings and Connectors
Chemical-resistant HDPE for use with Welch® Peristaltic Pump Systems (see 84009-696 series)
The barbed fittings kit 84009-610 includes four fittings each of all the tubing connectors for a total of 72 connectors.
No tools are required to connect tubing connectors to pumps.