Valor® 2000 Compact Industrial Bench Scale
Supplier: Ohaus
30035442 30035682 30035683 30035684 30035439 30035440 30035441 30035685 30251704 30251705
10123-878EA 1246.11 CAD
10123-878 10123-880 10123-882 10123-884 10123-886 10123-888 10123-890 10123-892 77434-820 77434-822
Valor® 2000 Compact Industrial Bench Scale
Balances and Scales
High-performance food scale suitable for harsh, wet workplace conditions. The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
  • IPX8 flow-thru design ensures that the NSF-certified Valor® 2000 will drain moisture in the scale when used in wet conditions
  • Stabilization time is less than a half-second
  • Large front and rear LED displays allow easy viewing so two operators can share one scale
The OHAUS Valor 2000 water resistant food scale raises the barMore Product Information