Valor® 3000 Xtreme Compact Scales, Ohaus
Supplier: Ohaus
83998130 83998132 83998133 83998135 83998136 83998134 83998131
97000-868EA 1787.07 CAD
97000-868 97000-872 97000-874 97000-876 97000-878 97000-880 97000-870
Valor® 3000 Xtreme Compact Scales, Ohaus
Balances and Scales
Valor™ scales combine high performance, ease of use, and portability into a rugged, full stainless steel design capable of operating in dry or wet applications
Standard features include software specifically designed for simple use and maximum throughput. Application modes include percentage weighing, specific gravity determination, and combined zero/tare, with results available in lb., decimal or fractional oz., lb.:oz., fluid oz., kg, or g units. Adjustable feet prevent slippage on wet or uneven surfaces, and can be set according to a front panel leveling indicator. An included AC adapter provides continuous power, and units will also run up to 100hrs. with four C-type batteries.
All Valor™ 3000 Xtreme models are USDA-AMS accepted, NSF certified/listed, and will support a HACCP certified system.
The compact and durable housing also offers chemical resistance and a removable weighing pan for simple cleaning. The advanced overload protection is rated at 10x capacity for demanding procedures, and units offer resolutions up to 20,000d for precise and repeatable measuring results. The intuitive user interface utilizes a bright backlit LCD and simple text prompts to guide users through setup and operation. Washdown versions ensure that the Valor™ 3000 Xtreme models will meet the needs of the most demanding environments.