VWR® Vacuum Pump Oil No.19
Supplier: VWR International
54996-106 54996-128 54996-061 54996-082
54996-106EA 682.18 CAD
54996-106 54996-128 54996-061 54996-082
VWR® Vacuum Pump Oil No.19
Lubricants
This is a superior oil based upon special crude oil stocks noted for their durability and fine lubricating qualities.
  • Durable
  • Fine lubricating qualities
  • Clean oil runs coolly in mechanical pumps
  • Improves system lifetime
The oil is further enhanced through precision distilling, resulting in a cleaner oil that runs cooler in most mechanical pumps. Cooler operation reduces thermal cracking for longer oil life. Good forepressures are prolonged, reducing backstreaming and there are often noticeable improvements in pumping cycle times.