Outlet Oil Mist Filters, Edwards
Supplier: Edwards
A46229000 A462-20-000 A46226000 A462-29-000 A462-26-000 A46220000
CAA462-20-000EA 921.52 CAD
EVA462-29-000 CAA462-20-000 CAA462-26-000 CAA462-29-000 EVA462-20-000 EVA462-26-000
Outlet Oil Mist Filters, Edwards
Filters Gas Filters
For efficient removal of oil vapors/odors from vacuum pump exhaust.
Ordering information: Complete with NW10 or NW25 adapters, NW clamp, and centering ring for easy installation.