Soldering Tips for PS-90 SensaTemp® Production Soldering Iron, PACE®
Supplier: Pace
1121-0305-P1 1121-0335-P5 1121-0337-P5 1121-0349-P5 1121-0358-P5 1121-0359-P5 1121-0361-P5 1121-0414-P5 1121-0499-P5 1121-0500-P5 1121-0527-P5 1121-0528-P5 1121-0529-P5 1121-0533-P5
30138-062EA 63.07 CAD
30138-062 30138-098 30138-102 30138-106 30138-114 30138-116 30138-118 30138-156 30138-200 30138-202 30138-224 30138-226 30138-228 30138-236
Soldering Tips for PS-90 SensaTemp® Production Soldering Iron, PACE®
Soldering Supplies Soldering Tips
Additional tips for PS-90 SensaTemp® Production Soldering Iron (30138-506)
SMT chip component tips remove resistor and capacity chip components; TSOP removal tips are ideal for thin, small outline package type components; PLCC removal tips remove PLCC sockets; SOT-23 removal tips remove small outline transistor type components; and PQFP removal tips remove PQFP type components.
Tips can also be used with PS-80, SP-2A, and IR-70 PACE® irons.
Tips feature a 0.48mm (3/16") shank diameter.