PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
56904SEAL12 56904SEAL18 44104SEL3000
52857-867EA 458.53 CAD
52857-867 52857-889 52857-903
PVC Film Wrap, T FRANK MCCALLS INC
Seals General Purpose Sealing Films
All-purpose, clear, polyvinyl wrap
Self-sealing. Adheres readily to glass vessels such as test tubes, flasks, cylinders, bottles, petri dishes, etc. Supplied in plain white carton.