96-Well Cap Mats, Greiner Bio-One
Supplier: Greiner Bio-One
381061 381070
82050-434CS 496.54 CAD
82050-434 82050-436 82050-440 82050-438
96-Well Cap Mats, Greiner Bio-One
Seals Microplate Seals
Cap mats are DMSO-resistant and suitable for applications requiring a removable sealer that is also exceptionally robust.
  • Ethyl vinyl acetate construction
  • Good temperature resistance
  • Alphanumeric coding for quick identification
  • Not pierceable
Cap mats for round wells are suitable for sealing 1 mL and 0.5 mL 96-well MASTERBLOCK® plates, in addition to standard 96-well polypropylene microplates
Cap mats for square wells are suitable for sealing 2 mL 96-well MASTERBLOCK® plates.
Cap mats are made of ethyl vinyl acetate (EVA) and can withstand temperatures from –20 to 60°C (–4 to 140°F). Alphanumerically labeled to ensure correct fitting and orientation.
The 96 well F-, U-, and µClear®-bottom polystyrene microplates may also be sealed (not suitable for 96-well V-bottom polystyrene microplates).
Packaging: Sterile cap mats are single packaged. Nonsterile cap mats are packaged 10 pieces per bag.