DuraSeal® Sealing Film, Diversified Biotech
Supplier: Diversified Biotech
DS2-500 DS4-250 DS4-500 DS5-150 DS12-250
89031-573CS 2812.62 CAD
89031-573 89031-575 89031-577 89031-579 10141-896
DuraSeal® Sealing Film, Diversified Biotech
Seals General Purpose Sealing Films
Durable, heat-resistant stretch sealing film is polyethylene-based and resists all caustic agents and organic solvents
DuraSharp® Shears are ideal for cutting DuraSeal® film. Shears are sold separately.
Allows containers to be incubated in boiling water baths without suffering the characteristic melting of paraffin-based alternatives. Film stretches symmetrically over a container and contracts inwardly around container opening or neck to provide a waterproof seal. Film can be elongated 500–800% before tearing.