VWR® Pierceable Films for ELISA and General Incubation
Supplier: VWR International
60941-088 60941-116
60941-088CS 145.06 CAD
60941-088 60941-116
VWR® Pierceable Films for ELISA and General Incubation
Seals Microplate Seals
These easily-pierceable 94.1 µm polyethylene films with acrylic adhesive allow direct sample recovery with single or multichannel pipettors and robotic probes.
  • For ELISA and general incubation applications
  • Functional temperature range from -40 to 90°C (-40 to 194°F)
  • Alphanumeric templates facilitate well location
Available with or without printed 96-well alphanumeric templates for easy well identification. The alphanumeric films include a white writing area for labeling and a color-coded dot on each end tab. Dimensions: 14.3x8.3 cm (55/8x31/4").
Ordering information: Color-coded films are supplied with an assortment of five colors, 10 films each.