Accessories for Microtiter® 96-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
Microtiter®
6604 9503130 5500 5701 5550 6305 3501
62402-929CS 477.33 CAD
62402-929 62403-072 62402-969 62402-936 62402-931 62402-921 53515-986
Accessories for Microtiter® 96-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
These accessories are for use with all Thermo Scientific Immulon®, Microfluor®, and Microlite™ Microtiter plates and Removawell* assemblies.
Rigid styrene lids are sterile. Flexible vinyl lids 62403-072 are for use with Removawell assemblies. Clear precut acetate, Mylar®, and PVC plate sealers are for storage and incubation procedures.
The Removawell holder 62402-969 holds eight 12-well or twelve 8-well Removawell strips in a 96-well format.