Pressure Tubes with Plunger Valve and Thermowell, Ace Glass
Supplier: Ace Glass
8648-164 8648-167 8648-170 8648-172 8648-175 8648-177 8648-179 8648-181
89063-252EA 277.99 CAD
89063-252 89063-258 89063-262 89063-264 89063-266 89063-268 89063-270 89063-272
Pressure Tubes with Plunger Valve and Thermowell, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are ideal for polymerization and analysis of halogens by thermal decomposition in the presence of lime
Available in a variety of lengths with #15 Ace-Threds. Tubes are pressure rated to 150psi at 120°C (320°F).
The unique Ace-Thred design allows for easy closing with supplied plug and O-ring. Closures are reusable. Tubes are equipped with bushing and plunger valve for purging. The glass plunger valve supplied with these tubes features a 5mm I.D. thermowell for a thermocouple wire or sheathed probe up to 3.2mm (1/8") O.D. The closed-bottom plungerMore Product Information