Super Erecta® Polymer Stem Casters, Metro™
Supplier: Metropolitan Wire
5PCR 5PC 5PCB
CA22221-206EA 301.1 CAD
CA22221-206 CA22233-712 CA22233-714
Super Erecta® Polymer Stem Casters, Metro™
Furniture Shelves
Use these stem casters with Super Erecta® shelving and SiteSelect® UP posts (see 22233-716 series) to create mobile units.
  • Innovative polymer stem casters offer corrosion-resistance and enhanced durability
  • Appropriate for all medium-duty mobile applications
Casters feature all-polymer horn, stainless steel axle and hardware, sleeve-style axle bearing and an optional toe-operated brake mechanism. They measure 12.7 cm (5") and are load rated at 135 kg (300 lbs.). Donut bumpers are included.