6000 Series Full Facepiece Respirators, 3M™
Supplier: 3M
7000002037 7000002029 7000002030
CA56612-880CS 1480.66 CAD
CA56612-880 CA56621-876 CA56621-878
6000 Series Full Facepiece Respirators, 3M™
Respirators Reusable Respirators Full Face Respirators
Reusable, lightweight full facepieces; respirators feature easy-to-adjust headstraps for maximum comfort. The Cool Flow® exhalation valve is designed to make breathing easier and to help reduce heat and moisture buildup in the facepiece.
  • Reusable, lightweight
  • Cool Flow® exhalation valve reduces heat and moisture buildup
  • Large lens is coated for scratch-resistance
  • NIOSH approved for negative pressure air purifying and positive pressure dual-airline applications
The center adapter directs exhaled breath and moisture downward
NIOSH approved for negative pressure air purifying and positive pressure dual airline applications.
Large lens is coated for scratch-resistance and provides excellent visibility. No retainers or extra parts are required to attach filter, cartridges, or dual airline. Respirators are also available with a DIN port adapter.
Accessories information: NIOSH approved cartridge protects against formaldehyde and certain organic vapors and can be used with bayonet filter holders.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.