KIMBLE® HI-VAC® Low Holdup Valves, Straight, with PTFE Plug, without Tip O-Ring, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
826620-0004 826620-0008
KT826620-0004EA 130.38 CAD
KT826620-0004 KT826620-0008
KIMBLE® HI-VAC® Low Holdup Valves, Straight, with PTFE Plug, without Tip O-Ring, Kimble Chase, DWK Life Sciences
Valves Vacuum Valves
Sidearms are bent to minimize holdup that normally occurs in conventional valves
PTFE shaft seals protect the Viton® O-rings from contact with the system. Seal design reduces outgassing.
Order Now
SPECIFICATIONS
Environmentally Preferable